
(2S,2′S)-2,2′-Bipyrrolidine

Synonym(s):
(S,S)-2,2′-Bipyrrolidine, (S,S)-2,2′-Bipyrrolidinyl
Empirical Formula (Hill Notation):
C8H16N2
CAS Number:
Molecular Weight:
140.23
Products
Brand
Formula Weight
Physical Form
Purity
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing