2-Hydroxy-2,2-di(2-thienyl)acetic acid
Synonym(s):
α-Hydroxy-α-2-thienyl-2-thiopheneacetic acid, 2-Hydroxy-2,2-di(thiophen-2-yl)acetic acid, Di-2-thienylglycolic acid
Empirical Formula (Hill Notation):
C10H8O3S2
Molecular Weight:
240.30
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing