Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Deoxy-D-ribose
Synonym(s):
2-Deoxy-D-arabinose, 2-Deoxy-D-erythropentose, Thyminose
Empirical Formula (Hill Notation):
C
5
H
10
O
4
CAS Number:
533-67-5
Molecular Weight:
134.13
EC Number:
208-573-0
Products
Product Category
carbohydrates (3)
monosaccharides (2)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Optical Isomer
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
121649
2-Deoxy-D-ribose,
97% (GC)
Expand
View Pricing
D5899
2-Deoxy-D-ribose,
BioReagent, suitable for cell culture
Expand
View Pricing
31170
2-Deoxy-D-ribose,
≥99.0% (TLC)
Expand
View Pricing