Skip to Content
Merck
2-Amino-6-bromobenzothiazole 97%

2-Amino-6-bromobenzothiazole

2-Amino-6-bromobenzothiazole 97%
Empirical Formula (Hill Notation):
C7H5BrN2S
CAS Number:
Molecular Weight:
229.10

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing