Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Amino-2-methyl-1,3-propanediol
Synonym(s):
AMPD, Ammediol, 2-Amino-2-methyl-1,3-propanediol
Linear Formula:
(HOCH
2
)
2
C(NH
2
)CH
3
CAS Number:
115-69-5
Molecular Weight:
105.14
EC Number:
204-100-7
Products
Product Category
buffers (2)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A9754
2-Amino-2-methyl-1,3-propanediol,
≥99%
Expand
View Pricing
8.01464
2-Amino-2-methyl-1,3-propanediol,
for synthesis
Expand
View Pricing
08569
2-Amino-2-methyl-1,3-propanediol,
BioUltra, ≥99.5% (NT)
Expand
View Pricing