Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,6-Pyridinedicarboxylic acid
Synonym(s):
DPA, DPAc, Dipicolinic acid
Empirical Formula (Hill Notation):
C
7
H
5
NO
4
CAS Number:
499-83-2
Molecular Weight:
167.12
EC Number:
207-894-3
Products
Product Category
acids (1)
building blocks (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P63808
2,6-Pyridinedicarboxylic acid,
99%
Expand
View Pricing
02321
2,6-Pyridinedicarboxylic acid,
suitable for ion chromatography, ≥99.5% (T)
Expand
View Pricing