Skip to Content
Merck
25-Hydroxyvitamin D3 solution 100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

25-Hydroxyvitamin D3 solution

25-Hydroxyvitamin D3 solution 100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade