2,2′-Oxydiethylamine dihydrochloride
Synonym(s):
2,2′-Oxybis(ethylamine) dihydrochloride, 2-Aminoethyl ether dihydrochloride
Linear Formula:
O(CH2CH2NH2)2 · 2HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing