
1-(tetrahydro-2-furoyl)-piperazine

Synonym(s):
(2RS)-1-(2-Tetrahydrofuroyl)piperazine
Empirical Formula (Hill Notation):
C9H16N2O2
CAS Number:
Molecular Weight:
184.24
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing