Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,3-Dimethyl-3,4,5,6-tetrahydro-2(1
H
)-pyrimidinone
Synonym(s):
1,3-Dimethyl-2-oxohexahydropyrimidine,
N,N′
-Dimethylpropylene urea, DMPU
Empirical Formula (Hill Notation):
C
6
H
12
N
2
O
CAS Number:
7226-23-5
Molecular Weight:
128.17
EC Number:
230-625-6
Products
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Absolute (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
251569
1,3-Dimethyl-3,4,5,6-tetrahydro-2(1
H
)-pyrimidinone,
98%, for peptide synthesis
Expand
View Pricing
41661
1,3-Dimethyl-3,4,5,6-tetrahydro-2(1
H
)-pyrimidinone,
absolute, over molecular sieve (H
2
O ≤0.05%), ≥99.0% (GC)
Expand
View Pricing