Skip to Content
Merck
1-(3-Chlorophenyl)piperazine hydrochloride 99%

1-(3-Chlorophenyl)piperazine hydrochloride

1-(3-Chlorophenyl)piperazine hydrochloride 99%
Synonym(s):
m-CPP
Empirical Formula (Hill Notation):
C10H13ClN2 · xHCl
CAS Number:
Molecular Weight:
196.68 (free base basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing