Skip to Content
Merck
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol ≥95% (TLC), liquid

1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol

1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol ≥95% (TLC), liquid
Synonym(s):
1,2-Di(cis-9,12-octadecadienoyl)-3-hexadecanoyl-rac-glycerol, PLL
Empirical Formula (Hill Notation):
C55H98O6
CAS Number:
Molecular Weight:
855.36

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing