MISSION® esiRNA
MISSION® esiRNA—powered by Eupheria Biotech—provide RNAi researchers with a proven, cost-effective, and simple way to perform post-transcriptional silencing of protein-coding genes and lncRNA (long non-coding RNA). Biologically-prepared, esiRNA are comprised of a heterogeneous pool of siRNA (natural RNA, no modifications) that all target the same mRNA sequence. These multiple silencing triggers lead to highly specific and effective gene knockdowns with lower off-target effects than single, chemically-synthesized siRNA.
To learn more about our MISSION® esiRNA products and their specifications, click here or search by gene target below.
Visit our Frequently Asked Questions page for more information.
Products
Related Product Resources
- Custom Oligos & qPCR Probes Online Tools
Learn more about the tools we offer for oligo and qpcr probe design, including OligoArchitect™ Primer and Probe Design Solutions and OligoEvaluator™: Oligonucleotide Sequence Calculator.
- Custom DNA/RNA Oligos & qPCR Probes Resources
Review resources for custom oligos, RNA oligos, and qPCR probes including online tools, case studies, and FAQs.
- Custom Oligo Blanket Orders
Custom oligo Blanket Purchase Order tracking system simplifies expenditure management for customers.
- RapidRED™ Payment Program
The payment program enables you to minimize paperwork and streamline your online ordering process with only one purchase transaction for setting up and depositing funds into your payment account.
- Oligo Product Reviews by SelectScience®
Custom & oredesigned DNA Oligos peer reviews. Check out what your peers are saying!