1699049
USP
Tropicamide Related Compound D
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Phenylacetic acid, α-Toluic acid, α-Tolylic acid, Benzeneacetic acid, PAA
About This Item
Recommended Products
grade
pharmaceutical primary standard
vapor density
~4 (vs air)
vapor pressure
1 mmHg ( 97 °C)
API family
tropicamide
manufacturer/tradename
USP
bp
265 °C (lit.)
mp
76-78 °C (lit.)
density
1.081 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
OC(=O)Cc1ccccc1
InChI
1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)
InChI key
WLJVXDMOQOGPHL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
269.6 °F - closed cup
Flash Point(C)
132 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service