Skip to Content
Merck
All Photos(1)

Documents

1609603

USP

Salmeterol xinafoate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±) 4-Hydroxy-a1-[[[6-(4-phenylbutoxy)hexyl]amino]m-ethyl]-1,3-benzenedimethanol xinafoate, GR 33343X xinafoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H37NO4 · C11H8O3
CAS Number:
Molecular Weight:
603.75
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

salmeterol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)c1ccc2ccccc2c1O.OCc3cc(ccc3O)C(O)CNCCCCCCOCCCCc4ccccc4

InChI

1S/C25H37NO4.C11H8O3/c27-20-23-18-22(13-14-24(23)28)25(29)19-26-15-7-1-2-8-16-30-17-9-6-12-21-10-4-3-5-11-21;12-10-8-4-2-1-3-7(8)5-6-9(10)11(13)14/h3-5,10-11,13-14,18,25-29H,1-2,6-9,12,15-17,19-20H2;1-6,12H,(H,13,14)

InChI key

XTZNCVSCVHTPAI-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salmeterol xinafoate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fluticasone Propionate and Salmeterol Inhalation Aerosol®
  • Fluticasone Propionate and Salmeterol Inhalation Powder
  • Salmeterol Inhalation Powder
  • Salmeterol Xinafoate

Biochem/physiol Actions

β2-adrenoceptor agonist.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Legal Information

Aerosol is a registered trademark of Cytec Technology Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R N Brogden et al.
Drugs, 42(5), 895-912 (1991-11-01)
Salmeterol xinafoate, like salbutamol (albuterol), is a saligenin derivative, and a selective beta 2-adrenoceptor agonist. It produces bronchodilation for at least 12 hours following inhalation of a single 50 micrograms dose. Salmeterol is intended for regular twice-daily treatment of reversible
A Jouyban-Gharamaleki et al.
International journal of pharmaceutics, 216(1-2), 33-41 (2001-03-29)
The mole fraction solubility of salmeterol xinafoate was determined in various concentrations of dioxane in aqueous binary mixture. Maximum solubility was observed in 90% v/v dioxane and solubility parameter of the solute was estimated from solubility peak equal to 24.99
R N Brogden et al.
Allergologia et immunopathologia, 20(2), 72-84 (1992-03-01)
Salmeterol xinafoate, like salbutamol (albuterol), is a saligenin derivative, and a selective beta 2-adrenoceptor agonist. It produces bronchodilation for at least 12 hours following inhalation of a single 50 micrograms dose. Salmeterol is intended for regular twice-daily treatment of reversible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service