Skip to Content
Merck
All Photos(1)

Key Documents

1089183

USP

Cannabinoids Mixture

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

API family

cannabis

manufacturer/tradename

USP

concentration

1 mL in methanol

storage temp.

−20°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cannabinoids Mixture USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Components

1 mL in Methanol:
  • Cannabichromene [CAS 20675-51-8]
  • Cannabidiol [CAS 13956-29-1]
  • Cannabidivarin [CAS 24274-48-4]
  • Cannabigerol [CAS 25654-31-3]
  • Cannabinol [CAS 521-35-7]
  • Delta-8-Tetrahydrocannabinol [CAS 5957-75-7]
  • Delta-9-Tetrahydrocannabinol [CAS 1972-08-3]
  • Tetrahydrocannabivarin [CAS 31262-37-0]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service