Skip to Content
Merck
All Photos(1)

Key Documents

93102

Supelco

Furocoumarin Mix

16 components in acetonitrile, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com


This product is suitable for the Furocoumarins quantification method developed by the IFRA.

Application


  • Bacterial Photomutagenicity Testing: Research on bacterial photomutagenicity testing utilized a furocoumarin mix to distinguish between direct, enzyme-mediated, and light-induced genetic alterations, highlighting its critical role in genetic toxicity studies (Utesch and Splittgerber, 1996).

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Psoralen
    5-Methoxypsoralen, phyproof® Reference Substance
    8-Methoxypsoralen
    Isopimpinellin
    Oxypeucedanin
    Oxypeucedanin hydrate
    Byakangelicin, phyproof® Reference Substance
    Biacangelicin
    (±)-Heraclenin
    8-geranyloxypsoralen
    Bergamottin
    Imperatorin
    Isoimperatorin
    Phellopterin
    6′,7′-Epoxybergamottin
    6´,-7´-Dihydroxybergamottin
    See All (16)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service