Skip to Content
Merck
All Photos(1)

Documents

505161

Supelco

Discovery® C18 (5 µm) HPLC Columns

L × I.D. 2 cm × 2.1 mm HPLC guard Column Kit with 1 Cartridge holder + 1 Guard Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 2.1 mm

Agency

suitable for USP L1

packaging

pkg of 1 kit

technique(s)

HPLC: suitable

L × I.D.

2 cm × 2.1 mm

matrix active group

C18 (octadecyl) bonding phase

particle size

5 μm

pore size

180 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Footnote

Kit includes one cartridge, a stand-alone holder, a piece of tubing, 2 nuts and ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T R Shantha Kumar et al.
Journal of pharmaceutical and biomedical analysis, 38(1), 173-179 (2005-05-24)
A simple, precise and accurate isocratic reverse-phase liquid chromatography method with programmed wavelength detection has been validated to quantify DRF-4367 and Phenol red, simultaneously for application in rat in situ single pass intestinal perfusion study to assess intestinal permeability of
Fabrice Gritti et al.
Journal of chromatography. A, 1095(1-2), 27-39 (2005-11-09)
The adsorption isotherms of an ionizable compound, nortriptyline, were accurately measured by frontal analysis (FA) on a C(18)-Discovery column, first without buffer (in an aqueous solution of acetonitrile at 15%, v/v of ACN), then with a buffer (in 28%, v/v
Ramisetti Nageswara Rao et al.
Journal of separation science, 32(9), 1312-1322 (2009-04-29)
Modafinil, adrafinil and their related substances were synthesized and analyzed by RP-LC with ESI-MS/MS. The ionization mode, polarity, cone voltage, and chromatographic conditions were evaluated. The optimum LC-MS conditions to obtain fragment ions indispensable for identification of the structures were
Himal Paudel Chhetri et al.
Saudi pharmaceutical journal : SPJ : the official publication of the Saudi Pharmaceutical Society, 22(5), 483-487 (2014-12-05)
This study presents the optimization of a simple HPLC-UV method for the determination of metformin in human plasma. Ion pair separation followed by UV detection was performed on deproteinized human plasma samples. The separation was carried out on a Discovery
Abbas Khan et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(24), 2423-2429 (2011-07-26)
A novel isocratic reversed-phase high performance liquid-chromatography/ultraviolet detection method for simultaneous determination of cefdinir and cefixime in human plasma was developed and validated after optimization of various chromatographic conditions and other experimental parameters. Sample preparation based on a simple extraction

Related Content

Discovery C18 and C8 HPLC Columns products offered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service