Skip to Content
Merck
All Photos(1)

Key Documents

40103-U

Supelco

δ-BHC

certified reference material, 1000 μg/mL in methanol

Synonym(s):

δ-HCH solution, δ-1,2,3,4,5,6-Hexachlorocyclohexane solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

ClC1C(C(C(C(C1Cl)Cl)Cl)Cl)Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H

InChI key

JLYXXMFPNIAWKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Wu et al.
Environmental microbiology, 9(9), 2331-2340 (2007-08-10)
Commercial formulations of hexachlorocyclohexane (HCH) consist of a mixture of four isomers, alpha, beta, gamma and delta. All these four isomers are toxic and recalcitrant pollutants. Sphingobium (formerly Sphingomonas) sp. strain BHC-A is able to degrade all four HCH isomers.
R Calvelo Pereira et al.
Environmental pollution (Barking, Essex : 1987), 155(2), 350-358 (2007-12-26)
This study focuses on the main routes of distribution and accumulation of different hexachlorocyclohexane (HCH) isomers (mainly alpha-, beta-, gamma- and delta-HCH) in a soil-plant-air system. A field assay was carried out with two plant species, Cynara scolymus L. and
Mrutyunjay Suar et al.
Applied and environmental microbiology, 70(11), 6650-6656 (2004-11-06)
Sphingomonas paucimobilis B90A is able to degrade the alpha-, beta-, gamma-, and delta-isomers of hexachlorocyclohexane (HCH). It contains the genes linA, linB, linC, linD, linE, and linR, which have been implicated in HCH degradation. In this study, dynamic expression of
Yong Qian et al.
Environmental monitoring and assessment, 116(1-3), 157-167 (2006-06-17)
The contamination of organochlorine pesticides hexachlorocyclohexane (HCH) and Dichlorodiphenyltrichloroethane (DDT) and their eco-environmental assessment in surface sediments from Lake Dongting, the second-largest freshwater lake in China, were studied. Concentrations of summation HCH (=alpha-HCH + beta-HCH + gamma-HCH +delta-HCH) were 0.21-9.59
Na Wang et al.
Chemosphere, 84(7), 964-971 (2011-07-05)
This paper presents a comprehensive study of pesticide levels and bio-accumulation characteristics in human adipose tissues among residents of Southeast China. A large number of adipose samples (n=633) were selected for 58 pesticides and were analyzed by high sensitive Gas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service