ZRB2550
Anti-CD66b/CEACAM8 Antibody, clone 1L6 ZooMAb® Rabbit Monoclonal
recombinant, expressed in HEK 293 cells
Synonym(s):
CD67 antigen, Carcinoembryonic antigen CGM6, Carcinoembryonic antigen-related cell adhesion molecule 8, Non-specific cross-reacting antigen NCA-95
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1L6, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 38.2 kDa
observed mol wt ~100 kDa
purified by
using Protein A
species reactivity
human
species reactivity (predicted by homology)
monkey
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal half
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... CEACAM8(1088)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in human peripheral blood mononuclear cells (PBMC) lysate.
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected CD66b/CEACAM8 in human peripheral blood mononuclear cells (PBMC) lysate.
Tested Applications
Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected CD66b/CEACAM8 in human spleen tissue.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD66b/CEACAM8 in Human peripheral blood mononuclear cells (PBMC) cells.
Flow Cytometry Analysis: 0.1 µg from a representative lot detected CD66b/CEACAM8 in human peripheral blood mononuclear cells (PBMC).
Affinity Binding Assay: The binding affinity of this antibody to CD66b/CEACAM8 peptide was at least one thousand-fold (1,000X) higher than that obtained with the non-specific control peptide.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service