Skip to Content
Merck
All Photos(1)

Key Documents

Z687502

3D Biotek 3D perfusion bioreactor system with pump

6 well chamber with PS scaffold inserts, autoclavable, AC/DC input 120 V AC

Synonym(s):

bioreactor systems, bioreactors, cell bioreactor, perfusion bioreactor, single use bioreactors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122105
NACRES:
NB.35

material

polycarbonate (4 interchangeable chambers)

sterility

non-sterile

feature

CSA approved
autoclavable
single-use bioreactor system

manufacturer/tradename

3D Biotek

W × D × H

14 in. × 13 in. × 13 in.

suitability

suitable for (stem cell growth and differentiation)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The 3D perfusion bioreactor consists of multiple independent, autoclavable polycarbonate chambers. The chambers are interchangeable and are specially designed to take advantage of 3D Insert scaffolds with various sizes ranging from 96-well to 6-well. Cell culture medias are perfused through the open porous structure of scaffolds using a pulsatile pump. The entire unit is autoclavable (except pump, of course) and can be used as a single-use bioreactor system.

Application

Numerous studies have proven that cells grow better under dynamic culture condition, simply because both nutrition supply and metabolic waste removal are better under dynamic culture conditions. Additionally, in some instances, the shear force produced by the flowing medium can act as a mechanical stimuli signal that will further promote stem cell differentiation toward certain cell lineage, such as osteoblast. Combined with the 3D Insert scaffold technology, the 3D Perfusion Bioreactor can facilitate the promotion of stem cell growth and differentiation, thereby enhancing stem cell research.

Legal Information

3D Insert is a trademark of 3D Biotek, LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service