Skip to Content
Merck
All Photos(1)

Key Documents

Z374873

PCR microtubes with attached caps

capacity 0.2 mL

Synonym(s):

PCR Tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.22

material

colorless cap
colorless tube
conical bottom
flat cap
polypropylene
polypropylene cap
polypropylene tube

description

Autoclavable

sterility

non-sterile

feature

RNase and DNase free
autoclavable at 121 °C
graduations: no
Push fit

packaging

pack of 250 ea
case of 4 packs

technique(s)

PCR: suitable

capacity

0.2 mL

color

clear

suitability

suitable for (PCR applications)

Looking for similar products? Visit Product Comparison Guide

General description

Thin-walled for efficient thermal transfer and shorter cycle times; fits all leading thermal cyclers including PerkinElmer, Biometra, MJ Research, Techne, Grant and Strategene (0.65 mL only). Certified DNase- and RNase-free. All tolerate organic solvent reactions and temperatures from -4 to 121 °C.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Reverse transcription (RT) is the process of converting RNA to cDNA using a reverse transcriptase enzyme and dNTPs.

Reverse transcription (RT) is the process of converting RNA to cDNA using a reverse transcriptase enzyme and dNTPs.

Reverse transcription (RT) is the process of converting RNA to cDNA using a reverse transcriptase enzyme and dNTPs.

Reverse transcription (RT) is the process of converting RNA to cDNA using a reverse transcriptase enzyme and dNTPs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service