Skip to Content
Merck
All Photos(2)

Key Documents

WH0004586M7

Sigma-Aldrich

Monoclonal Anti-MUC5AC antibody produced in mouse

clone 2H7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MUC5, Anti-mucin 5, subtypes A and C, tracheobronchial/gastric

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC5AC(4586)

General description

Mucin 5AC (MUC5AC) is encoded by the gene mapped to human chromosome 11p15.5 and is predominantly expressed in the gastric and tracheobronchial mucosae. The MUC5AC protein contains two types of deduced peptide domains such as eight amino acid tandemly repeated (TR) domains and cysteine-rich domains.

Immunogen

MUC5AC (XP_495860, 351 a.a. ~ 450 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
NQSTCAVYHRSLIIQQQGCSSSEPVRLAYCRGNCGDSSSMYSLEGNTVEHRCQCCQELRTSLRNVTLHCTDGSSRAFSYTEVEECGCMGRRCPAPGDTQH

Biochem/physiol Actions

Increased expression of Mucin 5AC (MUC5AC) leads to epithelial cancer progression, including colon cancer. Therefore, MUC5AC is considered to be a potential target in the treatment of colon cancer. MUC5AC containing the carbohydrate blood-group antigen, Lewis B (LeB) functions as a key receptor for Helicobacter pylori. O-glycoprotein encoded by the gene plays a vital role in mucus formation and epithelium protection. Decreased level of MUC5AC expression in ocular surface might lead to the tear instability in patients with SjÖgren syndrome.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pablo Argüeso et al.
Investigative ophthalmology & visual science, 43(4), 1004-1011 (2002-03-30)
To determine whether the relative amounts of mucin mRNA in the conjunctival epithelium and mucin protein in the tears are altered in patients with Sjögren syndrome compared with healthy individuals. Tear fluid was collected from the inferior fornix of normal
Erica Kosmerl et al.
Nutrients, 16(7) (2024-04-13)
The goblet cells of the gastrointestinal tract (GIT) produce glycoproteins called mucins that form a protective barrier from digestive contents and external stimuli. Recent evidence suggests that the milk fat globule membrane (MFGM) and its milk phospholipid component (MPL) can
V Guyonnet Duperat et al.
The Biochemical journal, 305 ( Pt 1), 211-219 (1995-01-01)
To date five human mucin cDNAs (MUC2, 5A, 5B, 5C and 6) mapped to 11p15.3-15.5, so it appears that this chromosome region might contain several distinct gene loci for mucins. Three of these cDNAs, MUC5A, B and C, were cloned
A E Biemer-Hüttmann et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 47(8), 1039-1048 (1999-07-29)
We studied the distribution of the four human apomucins MUC1, MUC2, MUC4, and MUC5AC in hyperplastic polyps, serrated adenomas, and traditional adenomas of the colorectum using immunohistochemical techniques, with the aim of comparing and contrasting their patterns of expression. A
Xijia Zhu et al.
Cancer biotherapy & radiopharmaceuticals, 31(7), 261-267 (2016-09-10)
Deregulated expressions of mucins have been found in various malignancies and play a pivotal role in carcinogenesis. MUC5AC, as a secreted mucin, is reported to be aberrantly expressed during epithelial cancer progression, including colon cancer. However, the mechanisms of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service