Skip to Content
Merck
All Photos(1)

Key Documents

U7508

Sigma-Aldrich

Anti-UVRAG antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-UV radiation resistance-associated gene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~90 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

western blot: 2-4 μg/mL using whole extracts of mouse brain and human Hela cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UVRAG(7405)
mouse ... Uvrag(78610)

Related Categories

General description

Ultraviolet irradiation resistance-associated gene (UVRAG) is an important autophagic tumor suppressor, encoded by the gene mapped to human chromosome 11q13.5. The encoded protein is characterized with four functional domains, such as proline-rich domain, a lipid-binding C2 domain, a beclin1-binding coiled-coil domain (CCD) and a C-terminal domain involved in centrosome integrity and DNA damage repair.

Immunogen

synthetic peptide corresponding to amino acids 665-678 of human UVRAG, conjugated to KLH. The corresponding sequence is identical in mouse.

Application

Anti-UVRAG antibody has been used in:
  • immunoblotting
  • western blotting
  • immunofluorescence

Anti-UVRAG antibody produced in rabbit has been used:
  • in western blotting
  • in immunofluorescence

Biochem/physiol Actions

Ultraviolet irradiation resistance-associated gene (UVRAG) is implicated in the regulation of intracellular membrane trafficking, including autophagy and chromosomal stability. The encoded protein regulates apoptosis by suppressing the BCL2-associated X protein (Bax) activity. Mutation in the gene has been observed in various types of human cancers, including microsatellite unstable colon carcinomas.

Target description

UVRAG activates the Beclin1-PI(3)KC3 complex, promoting autophagy and suppressing the proliferation and tumorigenicity of human colon cancer cells. This gene complements the ultraviolet sensitivity of xeroderma pigmentosum group C cells and encodes a prot

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UVRAG mutations associated with microsatellite unstable colon cancer do not affect autophagy
Knaevelsrud H, et al.
Autophagy, 6(7), 863-870 (2010)
Truncating mutation in the autophagy gene UVRAG confers oncogenic properties and chemosensitivity in colorectal cancers
He S
Nature Communications (2015)
UVRAG is required for organ rotation by regulating Notch endocytosis in Drosophila
Lee G, et al.
Developmental Biology, 356(2), 588-597 (2011)
Foot-and-mouth disease virus nonstructural protein 2C interacts with Beclin1, modulating virus replication
Gladue D P, et al.
Journal of Virology, 86(22), 12080-12090 (2012)
Chonsaeng Kim et al.
Journal of virology, 88(1), 434-443 (2013-10-25)
Echovirus 7 enters polarized Caco-2 intestinal epithelial cells by a clathrin-mediated endocytic process and then moves through the endosomal system before releasing its genome into the cytoplasm. We examined the possible role in virus entry of core components of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service