Skip to Content
Merck
All Photos(1)

Key Documents

T8575

Sigma-Aldrich

Anti-Tryptophan Hydroxylase antibody produced in sheep

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TPH

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

sheep

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

usage

sufficient for 10 blots

species reactivity

mammals, human

technique(s)

western blot: 1:1,000 using human dorsal raphe nucleus

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TPH1(7166)
mouse ... Tph1(21990)
rat ... Tph1(24848)

General description

The tryptophan hydroxylase 1 (TPH1) gene spanning 29kbp with 11 exons is mapped to human chromosome 11p15.3-p14. TPH1, is a 444 amino acid protein expressed in the gut, spleen, thymus and also in the pineal gland and the pituitary.
Tryptophan Hydroxylase (TPH) is a member of the aromatic amino acid hydroxylases (AAAH) family.

Immunogen

recombinant rabbit tryptophan hydroxylase, isolated as inclusion bodies from E. coli and purified by preparative SDS-PAGE.

Application

Anti-Tryptophan Hydroxylase antibody produced in sheep has been used in:
  • immunocytochemistry
  • immunohistochemistry
  • immunofluorescence
  • immunoperoxidase staining

Biochem/physiol Actions

The tryptophan hydroxylase 1 gene (TPH1) catalyzes the synthesis of 5-hydroxytryptophan, which is a precursor to the neurotransmitter serotonin. Variation in the gene expression affects prefrontal cortex activation during response inhibition. Mutation in the TPH1 gene leads to schizophrenia, irritable bowel syndrome (IBS) in women, suicidal behavior and depressive disorders. The encoded protein plays a vital role in regulation of cardiovascular function.

Physical form

Solution in 150 mM NaCl, 10 mM HEPES, pH 7.5, 100 μg per mL BSA, and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew W Hale et al.
Journal of chemical neuroanatomy, 41(4), 219-226 (2011-06-07)
Tryptophan hydroxylase is the rate-limiting enzyme in the biosynthesis of serotonin (5-hydroxytryptamine; 5-HT). Two isoforms of tryptophan hydroxylase, derived from different genes, tph1 and tph2, have been identified. The tph1 isoform is expressed in peripheral tissues, whereas tph2 is brain
Tryptophan hydroxylase 1 gene polymorphisms alter prefrontal cortex activation during response inhibition.
Ruocco AC
Neuropsychology, 30, 18-27 (2016)
R C Drugan et al.
Neuroscience, 253, 221-234 (2013-09-04)
Prior adverse experience alters behavioral responses to subsequent stressors. For example, exposure to a brief swim increases immobility in a subsequent swim test 24h later. In order to determine if qualitative differences (e.g. 19°C versus 25°C) in an initial stressor
GABAergic neurons in the dorsal raphe nucleus that express 5-HT3A receptors participate in responses to stress hormones
Li X, et al.
Neuroscience, 9(7), e101420-e101420 (2020)
Matthew W Hale et al.
Experimental neurology, 224(1), 271-281 (2010-04-13)
Serotonin plays an important role in the regulation of anxiety states and physiological responses to aversive stimuli. Intracerebroventricular (i.c.v.) injection of the stress- and anxiety-related neuropeptide urocortin 2 (Ucn 2) increases c-Fos expression in serotonergic neurons in the dorsal (DRD)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service