Skip to Content
Merck
All Photos(1)

Key Documents

T6901

Sigma-Aldrich

α1-Mating Factor acetate salt

≥93% (HPLC), synthetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C82H114N20O17S
CAS Number:
Molecular Weight:
1683.97
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic

Quality Level

Assay

≥93% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C1CCCN1C(=O)C(CCC(N)=O)NC(=O)CNC(=O)C2CCCN2C(=O)C(CCCCN)NC(=O)C(CC(C)C)NC(=O)C(CCC(N)=O)NC(=O)C(CC(C)C)NC(=O)C(Cc3c[nH]c4ccccc34)NC(=O)C(Cc5cnc[nH]5)NC(=O)C(N)Cc6c[nH]c7ccccc67)C(=O)NC(Cc8ccc(O)cc8)C(O)=O

InChI

1S/C82H114N20O17S/c1-45(2)34-61(98-76(112)63(38-49-41-89-56-17-9-7-15-53(49)56)99-77(113)64(39-50-42-87-44-91-50)96-71(107)54(84)37-48-40-88-55-16-8-6-14-52(48)55)74(110)93-57(25-27-68(85)104)72(108)97-62(35-46(3)4)75(111)95-59(18-10-11-30-83)80(116)101-31-12-19-66(101)78(114)90-43-70(106)92-60(26-28-69(86)105)81(117)102-32-13-20-67(102)79(115)94-58(29-33-120-5)73(109)100-65(82(118)119)36-47-21-23-51(103)24-22-47/h6-9,14-17,21-24,40-42,44-46,54,57-67,88-89,103H,10-13,18-20,25-39,43,83-84H2,1-5H3,(H2,85,104)(H2,86,105)(H,87,91)(H,90,114)(H,92,106)(H,93,110)(H,94,115)(H,95,111)(H,96,107)(H,97,108)(H,98,112)(H,99,113)(H,100,109)(H,118,119)

InChI key

SBKVPJHMSUXZTA-UHFFFAOYSA-N

Amino Acid Sequence

Trp-His-Trp-Leu-Gln-Leu-Lys-Pro-Gly-Gln-Pro-Met-Tyr

Application

α1-Mating Factor acetate salt has been used for the synchronization of yeast cells for flow cytometry and to study B-type cyclin (Clb2p)-EmCitrine levels from calcofluor white treated cells.

Biochem/physiol Actions

The α1-Mating factor is a pheromone produced by yeast that regulates mating. α1-Mating Factor acetate salt can be utilized to arrest the G1-phase of the cell cycle in various yeast strains.

Preparation Note

Alpha1-Mating Factor acetate salt dissolves in water at 1 mg/ml to yield a clear, colorless solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Formosa et al.
Journal of molecular recognition : JMR, 28(1), 1-9 (2015-06-06)
Single-molecule force spectroscopy using atomic force microscopy (AFM) is more and more used to detect and map receptors, enzymes, adhesins, or any other molecules at the surface of living cells. To be specific, this technique requires antibodies or ligands covalently
Brianna L Greenwood et al.
Methods in molecular biology (Clifton, N.J.), 2579, 145-168 (2022-09-01)
The cell division cycle is a fundamental process required for proliferation of all living organisms. The eukaryotic cell cycle follows a basic template with an ordered series of events beginning with G1 (Gap1) phase, followed successively by S (Synthesis) phase
Oliver Kerscher et al.
Molecular and cellular biology, 23(18), 6406-6418 (2003-08-29)
Spindle checkpoint proteins monitor the interaction of the spindle apparatus with the kinetochores, halting anaphase even if the microtubule attachment of only a single chromosome is altered. In this study, we show that Bub3p of Saccharomyces cerevisiae, an evolutionarily conserved
Jonay García-Luis et al.
Genes, 9(11) (2018-11-21)
Homologous recombination (HR) is a preferred mechanism to deal with DNA replication impairments. However, HR synapsis gives rise to joint molecules (JMs) between the nascent sister chromatids, challenging chromosome segregation in anaphase. Joint molecules are resolved by the actions of
Stefano Vernarecci et al.
Molecular and cellular biology, 28(3), 988-996 (2007-11-28)
We report that the histone acetyltransferase Gcn5p is involved in cell cycle progression, whereas its absence induces several mitotic defects, including inefficient nuclear division, chromosome loss, delayed G(2) progression, and spindle elongation. The fidelity of chromosome segregation is finely regulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service