Skip to Content
Merck
All Photos(1)

Documents

T6634

Sigma-Aldrich

Thrombin from bovine plasma

lyophilized powder, 600-2,000 NIH units/mg protein (biuret)

Synonym(s):

Factor IIa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

600-2,000 NIH units/mg protein (biuret)

mol wt

heavy chain ~33 kDa
light chain ~5 kDa

UniProt accession no.

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Gene Information

cow ... F2(280685)

Looking for similar products? Visit Product Comparison Guide

General description

Thrombin is a sodium-activated type II enzyme. It contains two anion binding exosites, ABE-I and ABE-II. The predominant form of thrombin in vivo is the zymogen prothrombin (factor II), which is produced in the liver. Bovine a-thrombin consists of a light chain (A chain) and a heavy chain (B chain). These two chains are joined by one disulfide bond. The B chain of a-thrombin includes a carbohydrate portion.

Application

Thrombin from bovine plasma has been used to study its effect on the perinatal rat subventricular zone cells and oligodendrocyte precursor cell proliferation, differentiation, and migration in culture. It has also been used in fibrin degradation assay to measure nattokinase activity.
Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags. Thrombin has been used in a study to assess global haemostasis and point of care testing.

Biochem/physiol Actions

Thrombin is a proteolytic enzyme critical in the blood clotting process and activates clotting factors V, VIII, XI, and XII. Thrombin promotes platelet aggregation. Therefore, thrombin is the final coagulation protease in hemostasis, promoting both procoagulant and anticoagulant effects. It is used to treat bleeding from capillaries and small venules.
Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Unit Definition

Activity is expressed in NIH units obtained by direct comparison to a NIH thrombin reference standard.

Physical form

Lyophilized from saline sodium citrate buffer, pH 6.5

Analysis Note

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard, Lot K.
The NIH assay procedure uses 0.2 mL of diluted plasma (1:1 with saline) as a substrate and 0.1 mL of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Other Notes

View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thrombin as an Agent
xPharm: The Comprehensive Pharmacology Reference null
Thrombin as a Target
xPharm: The Comprehensive Pharmacology Reference null
Y Dargaud et al.
Haemophilia : the official journal of the World Federation of Hemophilia, 18 Suppl 4, 81-88 (2012-06-29)
The evaluation of the coagulation profile has used so far either clotting-based or chromogenic assays with different endpoints. Clotting-based techniques are the most used worldwide, and they certainly are useful for diagnosis of clotting factor deficiencies. However, the information provided
Congenital and Acquired Hypercoagulable Syndromes
The Vein Book, 339-346 (2007)
Hongkai Xiang et al.
The Journal of international medical research, 48(9), 300060520957541-300060520957541 (2020-09-26)
To assess changes in plasma exosome levels and protein content in mice after long-term exercise. We subjected 9-month-old adult C57BL/6J mice to daily treadmill running exercise for 4 weeks prior to the isolation of blood-derived exosomes. Exosomal proteins were identified

Articles

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service