Skip to Content
Merck
All Photos(1)

Documents

T3309

Sigma-Aldrich

Transferrin human

≥98%

Synonym(s):

Siderophilin, partially saturated

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human

Quality Level

Assay

≥98%

form

powder

Iron content

300-600 ppm

technique(s)

cell culture | stem cell: suitable

impurities

≤1.0 EU/mg endotoxin

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... TF(7018)

Looking for similar products? Visit Product Comparison Guide

General description

Transferrin is an 80 kDa serum glycoprotein, which has an half-life of 8 to 10 days. The gene is located on human chromosome 3q22.1. Transferrin is expressed in hepatocytes.

Application

Transferrin has been used:
  • in isolation of epidermal stem cells and cell expansion
  • to study the effect of compound treatment on transferrin uptake
  • for transferrin uptake and confocal imaging

Biochem/physiol Actions

Transferrin facilitates the transfer of iron to erythron and peripheral tissues. Mutations in this gene is associated with atransferrinemia. Transferrin is required for fetal development. It is non immunogenic and transferrin-conjugated drugs inhibits cardiotoxicity and drug resistance.
Transferrin was shown to promote the proliferation of human lymphocytes. This effect is directly proportional to the level of iron present.

Analysis Note

Purity by agarose electrophoresis.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TFRC (transferrin receptor (p90, CD71))
Dorak MT
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2009)
Nutritional issues in the patient with diabetes and foot ulcers
LITCHFORD MD
Levin and O'Neal's The Diabetic Foot, 199-217 (2008)
A Djeha et al.
British journal of haematology, 80(2), 235-241 (1992-02-01)
The effect of different forms of iron and iron-binding proteins on the proliferative response of human lymphocytes to phytohaemagglutinin (PHA) has been studied. Transferrin enhanced proliferation, the effect being proportional to the degree of iron saturation up to 100%, but
Non-invasive in vivo imaging of near infrared-labeled transferrin in breast cancer cells and tumors using fluorescence lifetime FRET
Abe K, et al.
PLoS ONE, 8(11), e80269-e80269 (2013)
Large scale screening and identification of novel Ebolavirus and Marburgvirus entry inhibitors
Anantpadma M, et al.
Antimicrobial Agents and Chemotherapy, AAC-00543 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service