Skip to Content
Merck
All Photos(3)

Key Documents

T3134

Sigma-Aldrich

N,N,N′,N′-Tetramethyl-p-phenylenediamine dihydrochloride

≥95%, powder

Synonym(s):

TMPPD, Wurster’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4[N(CH3)2]2·2HCl
CAS Number:
Molecular Weight:
237.17
Beilstein:
7442241
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥95%

form

powder

color

off-white to gray-brown

mp

222-224 °C (lit.)

solubility

H2O: 500 mg/10 mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].Cl[H].CN(C)c1ccc(cc1)N(C)C

InChI

1S/C10H16N2.2ClH/c1-11(2)9-5-7-10(8-6-9)12(3)4;;/h5-8H,1-4H3;2*1H

InChI key

FBHKTSXMTASXFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N,N′,N′-Tetramethyl-p-phenylenediamine dihydrochloride (TMPD) is a redox mediator used in the oxidase test for detecting bacterial cytochrome c oxidases. TMPD is commonly used to perform the oxidase test for the colorimetric identification of pathogenic and non-pathogenic bacteria.

Application

N,N,N′,N′-Tetramethyl-p-phenylenediamine dihydrochloride has been used to determine the oxidase activity displayed by various microorganisms.

Caution

May darken in storage.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Leucobacter chironomi sp. nov., a chromate-resistant bacterium isolated from a chironomid egg mass
Malka H
International Journal of Systematic and Evolutionary Microbiology (2009)
Siamak Heidari Nonakaran et al.
World journal of microbiology & biotechnology, 31(12), 1993-2001 (2015-09-21)
Biomineralization is a process by which living organisms produce minerals. The extracellular production of these biominerals by microbes has potential for various bioengineering applications. For example, crack remediation and improvement of durability of concrete is an important goal for engineers
Paenibacillus sacheonensis sp. nov., a xylanolytic and cellulolytic bacterium isolated from tidal flat sediment
Jeong C
International Journal of Systematic and Evolutionary Microbiology (2011)
Yen-Chi Tsao et al.
International journal of molecular sciences, 21(12) (2020-06-25)
Isoplumbagin (5-hydroxy-3-methyl-1,4-naphthoquinone), a naturally occurring quinone from Lawsonia inermis and Plumbago europaea, has been reported to have anti-inflammatory and antimicrobial activity. Inflammation has long been implicated in cancer progression. In this study, we examined the anticancer effect of chemically synthesized
Meng Gao et al.
Journal of cellular physiology, 236(4), 2920-2933 (2020-09-16)
The previous research has shown that mitochondrial flash (mitoflash) genesis are functionally and mechanistically integrated with mitochondrial electron transport chain (ETC) energy metabolism. However, the response of mitoflash to superoxide is not entirely consistent with the response of MitoSOX Red.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service