Skip to Content
Merck
All Photos(1)

Key Documents

T1573

Sigma-Aldrich

Thrombin Receptor Activator Peptide 6

>95% (HPLC)

Synonym(s):

TRAP-6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H56N10O9
CAS Number:
Molecular Weight:
748.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>95% (HPLC)

form

lyophilized

technique(s)

cell culture | mammalian: suitable

solubility

0.1% TFA: soluble
water: soluble

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)CO)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(N)=O)C(O)=O

InChI

1S/C34H56N10O9/c1-18(2)13-23(30(49)40-22(11-8-12-39-34(37)38)29(48)44-26(33(52)53)16-27(36)46)42-31(50)24(14-19(3)4)43-32(51)25(41-28(47)21(35)17-45)15-20-9-6-5-7-10-20/h5-7,9-10,18-19,21-26,45H,8,11-17,35H2,1-4H3,(H2,36,46)(H,40,49)(H,41,47)(H,42,50)(H,43,51)(H,44,48)(H,52,53)(H4,37,38,39)/t21-,22-,23-,24-,25-,26-/m0/s1

InChI key

HAGOWCONESKMDW-FRSCJGFNSA-N

Gene Information

human ... F2R(2149)

Related Categories

Amino Acid Sequence

Ser-Phe-Leu-Leu-Arg-Asn

General description

Thrombin Receptor Activator Peptide 6 (TRAP-6) protects astrocytes from cell death, which is associated with hypoglycemia. TRAP-6 induced aggregation and impaired adhesion to collagen under flow conditions.

Application

Thrombin Receptor Activator Peptide 6 has been used to activate platelets in vitro. It has also been used to downregulate hippo signaling and increase nuclear yes-associated protein (YAP).

Biochem/physiol Actions

Hexapeptide fragment of the thrombin receptor (residues 42-47).
Hexapeptide fragment of the thrombin receptor (residues 42-47). Shown to activate the thrombin receptor and cause platelet aggregation (EC50 = 0.8 μM).

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xavier Charest-Morin et al.
Frontiers in immunology, 9, 2183-2183 (2018-10-20)
Multiple pathways have been proposed to generate bradykinin (BK)-related peptides from blood. We applied various forms of activation to fresh blood obtained from 10 healthy subjects or 10 patients with hereditary angioedema (HAE-1 or -2 only) to investigate kinin formation.
Saskia Wand et al.
Clinical and applied thrombosis/hemostasis : official journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis, 24(1), 179-185 (2017-03-18)
We aimed to identify the prevalence of acetylsalicylic acid (ASA) nonresponse in patients after coronary artery bypass graft (CABG) surgery and the possible consequences for the rate of major cardiovascular events. This prospective, observational, bicentric cohort study was conducted in
JCAD, a gene at the 10p11 coronary artery disease locus, regulates Hippo signaling in endothelial cells
Jones PD, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 38(8), 1711-1722 (2018)
Hematology: Basic Principles and Practice E-Book: Basic Principles and Practice (2017)
Kristoffer Navred et al.
PloS one, 14(1), e0211130-e0211130 (2019-01-24)
Flow cytometric platelet activation has emerged as an alternative diagnostic test for inherited platelet disorders. It is, however, labor intensive and few studies have directly compared the performance of flow cytometric platelet activation (PACT) to light transmission aggregometry (LTA). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service