Skip to Content
Merck
All Photos(1)

Documents

T1452

Sigma-Aldrich

TPCA-1

≥95% (HPLC)

Synonym(s):

[5-(p-Fluorophenyl)-2-ureido]thiophene-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10FN3O2S
CAS Number:
Molecular Weight:
279.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

NC(=O)Nc1sc(cc1C(N)=O)-c2ccc(F)cc2

InChI

1S/C12H10FN3O2S/c13-7-3-1-6(2-4-7)9-5-8(10(14)17)11(19-9)16-12(15)18/h1-5H,(H2,14,17)(H3,15,16,18)

InChI key

SAYGKHKXGCPTLX-UHFFFAOYSA-N

Application

TPCA-1 has been used to study its cytotoxic effects in RPMI8226 and LP1 cell lines. It has also been used as a culture media supplement for nuclear protein fractions extracted from human airway smooth muscle cells.

Biochem/physiol Actions

TPCA-1 is a potent and selective inhibitor of human IκB kinase-2 (IKK-2) with IC50 = 17.9 nM for IKK-2 compared to 400nm for IKK-1. It has been used to study inhibition of IKK-2 to prevent inflammatory mediator release in animal models of arthritis and airway inflammation.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

TPCA-1 is soluble in DMSO at a concentration that is greater than or equal to 10 mg/ml. It is insoluble in water.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ines Tagoug et al.
PloS one, 6(7), e22641-e22641 (2011-07-30)
Multiple myeloma (MM) is a B cell neoplasm characterized by bone marrow infiltration with malignant plasma cells. IGF-1 signalling has been explored as a therapeutic target in this disease. We analyzed the effect of the IKK2 inhibitor AS602868, in combination
Lauren D Aarreberg et al.
mBio, 9(2) (2018-03-22)
Induction of interferon beta (IFN-β), IFN-stimulated genes (ISGs), and inflammatory responses is critical for control of viral infection. We recently identified an essential linkage of stimulation of the inflammatory cytokine interleukin-1β (IL-1β) and induction of ISGs that function as host
Alice Borghi et al.
Biochemical pharmacology, 153, 292-298 (2018-01-30)
Tumor Necrosis Factor (TNF) is a proinflammatory cytokine that elicits its action by binding to two cell surface TNF receptors (TNFR), TNFR1 and TNFR2, which are expressed by many different cell types. Stimulation of TNFR1 activates canonical NF-κB signaling, leading
Galangin Abrogates Ovalbumin-Induced Airway Inflammation via Negative Regulation of NF-B.
Zha, W. J., et al.
Evidence-Based Complementary and Alternative Medicine : ECAM, 2013, doi:10-doi:10 (2013)
Mark A Birrell et al.
Molecular pharmacology, 69(6), 1791-1800 (2006-03-07)
Nuclear factor kappaB (NF-kappaB) is a transcription factor believed to be central in the expression of numerous inflammatory genes and the pathogenesis of many respiratory diseases. We have previously demonstrated increased NF-kappaB pathway activation in a steroid-sensitive animal model of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service