Key Documents
T1418
Sigma-Aldrich
Nalgene® centrifuge tubes, Oak Ridge Style 3119
nominal capacity 50 mL
Synonym(s):
High-Speed ​​Centrifuge Tubes
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
polypropylene copolymer
polypropylene screw closure
round bottom tube
screw top cap (Linerless)
sterility
non-sterile
feature
RCF 100,605 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3119-0050
nominal capacity
50 mL
General description
“Nalgene” Oak Ridge high-speed polypropylene copolymer (PPCO) centrifuge tubes offer broader chemical compatibility with lab reagents compared to polycarbonate centrifuge tubes.
Features and Benefits
  • Oak Ridge style centrifuge tubes are strong with excellent mechanical strength.
  • Chemically resistant and can withstand repeated autoclave cycles.
  • Clear tubes allow for optimal sample visibility.
  • Polypropylene screw closure provides a leak-proof seal to prevent sample loss.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries