Skip to Content
Merck
All Photos(4)

Key Documents

T0665

Sigma-Aldrich

holo-Transferrin human

powder, BioReagent, suitable for cell culture, ≥97%

Synonym(s):

Siderophilin, iron-saturated

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

biological source

human plasma

Quality Level

description

Iron-saturated

product line

BioReagent

Assay

≥97%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

HIV, HBsAg and HCV, none detected (tested by FDA approved testing methods)
endotoxin, tested

solubility

H2O: 50 mg/mL

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... TF(7018)

Looking for similar products? Visit Product Comparison Guide

Application

Transferrin is the primary iron binding and cell delivery molecule present in serum. Serum-free cell culture systems require a delivery format for iron. Transferrin is the preferential delivery form of iron because cells process transferrin bound iron in a physiologically appropriate way through transferrin receptors on the cell surface. Human holo-transferrin is a high affinity transferrin that can be used with a wide range of cells types. Human holo-transferrin is loaded with iron prior to use and can be added directly to iron poor or iron-free cell culture media.
holo-Transferrin human has been supplemented in Roswell Park Memorial Institute (RPMI) for proerythroblasts proliferation assay and also in culture media for inductively coupled plasma emission spectroscopy.

Biochem/physiol Actions

Provides iron to cells in a physiologically stable and safe form.

Preparation Note

Heat treated at 60 °C (minimum) for a minimum of 10 hours.

Analysis Note

Iron content by ICP reported on a lot-specific basis.
Purity by agarose gel electrophoresis.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jill M Brown et al.
Nature communications, 9(1), 3849-3849 (2018-09-23)
Self-interacting chromatin domains encompass genes and their cis-regulatory elements; however, the three-dimensional form a domain takes, whether this relies on enhancer-promoter interactions, and the processes necessary to mediate the formation and maintenance of such domains, remain unclear. To examine these
Co-operative signalling mechanisms required for erythroid precursor expansion in response to erythropoietin and stem cell factor
Arcasoy O and Jiang X
British Journal of Haematology, 130(1), 121-129 (2005)
Engineered mitochondrial ferritin as a magnetic resonance imaging reporter in mouse olfactory epithelium
Iordanova B, et al.
Testing, 8(8), e72720-e72720 (2013)
Pamela Sarkar et al.
Cytotherapy, 20(1), 21-28 (2017-09-18)
Clinical trials using ex vivo expansion of autologous mesenchymal stromal cells (MSCs) are in progress for several neurological diseases including multiple sclerosis (MS). Given that environment alters MSC function, we examined whether in vitro expansion, increasing donor age and progressive
Senthil Raja Jayapal et al.
Cell cycle (Georgetown, Tex.), 15(22), 3070-3081 (2016-09-23)
Cyclin A2 is an essential gene for development and in haematopoietic stem cells and therefore its functions in definitive erythropoiesis have not been investigated. We have ablated cyclin A2 in committed erythroid progenitors in vivo using erythropoietin receptor promoter-driven Cre

Protocols

Discover our primary human hepatocytes and plating and maintenance media formulations. Explore our protocol for plateable primary hepatocyte cell culture.

Discover our primary human hepatocytes and plating and maintenance media formulations. Explore our protocol for plateable primary hepatocyte cell culture.

Discover our primary human hepatocytes and plating and maintenance media formulations. Explore our protocol for plateable primary hepatocyte cell culture.

Discover our primary human hepatocytes and plating and maintenance media formulations. Explore our protocol for plateable primary hepatocyte cell culture.

See All

Related Content

Explore our protocol for suspension primary hepatocyte cell culture. Discover our primary human hepatocytes and plating and maintenance media formulations.

Explore our protocol for suspension primary hepatocyte cell culture. Discover our primary human hepatocytes and plating and maintenance media formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service