Skip to Content
Merck
All Photos(1)

Key Documents

T0578

Sigma-Aldrich

Teicoplanin

Synonym(s):

Targocid, Teichomycin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
51282701
NACRES:
NA.76

Assay

≥80% (teicoplanins A2, HPLC)

Quality Level

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

white to faint beige
white to faint yellow
white to light brown

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

InChI

1S/C88H97Cl2N9O33/c1-3-4-5-6-7-8-9-10-60(108)94-68-74(113)71(110)58(32-101)129-87(68)132-78-55-26-40-27-56(78)126-52-18-14-38(24-47(52)90)77(131-86-67(92-34(2)103)73(112)70(109)57(31-100)128-86)69-84(121)98-66(85(122)123)45-29-42(105)30-54(127-88-76(115)75(114)72(111)59(33-102)130-88)61(45)44-23-37(13-15-49(44)106)63(81(118)99-69)96-83(120)65(40)97-82(119)64-39-21-41(104)28-43(22-39)124-53-25-36(12-16-50(53)107)62(91)80(117)93-48(79(116)95-64)20-35-11-17-51(125-55)46(89)19-35/h11-19,21-30,48,57-59,62-77,86-88,100-102,104-107,109-115H,3-10,20,31-33,91H2,1-2H3,(H,92,103)(H,93,117)(H,94,108)(H,95,116)(H,96,120)(H,97,119)(H,98,121)(H,99,118)(H,122,123)/t48-,57-,58-,59-,62-,63-,64+,65-,66-,67-,68-,69+,70-,71-,72-,73-,74-,75+,76+,77-,86+,87+,88+/m1/s1

InChI key

BJNLLBUOHPVGFT-QRZIFLFXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Teicoplanin is a glycopeptide antibiotic produced by Actinoplanes teichomiceticus that is effective against Gram-positive bacteria resistant to β-lactam antibiotics. Teicoplanin and vancomycin have been used clinically for the treatment of methicillin-resistant Staphylococcus aureus (MRSA) infections. Product T0578 is a mixture of six primary teicoplanins; primarily teicoplanins A2 group.

Biochem/physiol Actions

Teicoplanin is a glycopeptide antibiotic. It is a mixture of several compounds, five major (named teicoplanin A2-1 through A2-5) and four minor (named teicoplanin RS-1 through RS-4). All teicoplanins share a same glycopeptide core, termed teicoplanin A3-1, a fused ring structure to which two carbohydrates (mannose and N-acetylglucosamine) are attached. The major and minor components also contain a third carbohydrate moiety, β-D-glucosamine, and differ only by the length and conformation of a side chain attached to it. It′s mechanism of action is through inhibition of bacterial cell wall biosynthesis.
Teicoplanin is a glycopeptide antibiotic produced by Actinoplanes teichomiceticus that is effective against Gram-positive bacteria resistant to β-lactam antibiotics. Its mechanism of action is through inhibition of bacterial cell wall biosynthesis.

Packaging

100MG,1G

Other Notes

Mixture of six primary teicoplanins; primarily teicoplanins A2 group.
Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annamaria Iagnocco et al.
Medical ultrasonography, 14(3), 251-253 (2012-09-08)
Musculoskeletal ultrasound (US) is gaining an increasing role in the assessment of a variegate set of joint and periarticular soft tissues abnormalities in rheumatology. In addition, US represents a useful technique in guiding local procedures such as aspiration of fluid
Szilvia Tollas et al.
European journal of medicinal chemistry, 54, 943-948 (2012-07-17)
Glycopeptide antibiotic derivative teicoplanin ψ-aglycone has been bound covalently to a fullerenopyrrolidine derivative using azide-alkyne 1,3-dipolar cycloaddition reaction. The aggregation of the antibiotic-fullerene conjugate in aqueous solution has been studied. The conjugate exhibited antibacterial activity against enterococci resistant to teicoplanin.
Jann-Tay Wang et al.
BMC infectious diseases, 13, 182-182 (2013-04-23)
The present study was designed to investigate whether teicoplanin minimum inhibitory concentrations (MICs) of methicillin-resistant Staphylococcus aureus (MRSA) isolates play a role in the prognosis of patient with teicoplanin-treated MRSA bloodstream infection (BSI). Between 1 January 2006 and 31 December
Hongli Sun et al.
Diagnostic microbiology and infectious disease, 74(4), 399-403 (2012-10-27)
The epidemiology and molecular characteristics of vancomycin-resistant enterococci (VRE) from China deserve further investigation. This study reports on the molecular characterization of 101 unique VRE (96 E. faecium and 5 E. faecalis strains) recovered from diverse samples of 12 hospitals
Jinjian Fu et al.
PloS one, 8(3), e58240-e58240 (2013-03-14)
To assess the effectiveness and safety of linezolid in comparison with glycopeptides (vancomycin and teicoplanin) for the treatment of Staphylococcus aureus infections, we conducted a meta-analysis of relevant randomized controlled trials. A thorough search of Pubmed and other databases was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service