Skip to Content
Merck
All Photos(2)

Key Documents

T0535

Sigma-Aldrich

Tolfenamic acid

NSAID

Synonym(s):

2 (3-Chloro-2-methylanilino)benzoic acid, 2-([3-Chloro-2-methylphenyl]amino)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12ClNO2
CAS Number:
Molecular Weight:
261.70
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

ethanol: 50 mg/mL, clear, greenish-yellow

SMILES string

Cc1c(Cl)cccc1Nc2ccccc2C(O)=O

InChI

1S/C14H12ClNO2/c1-9-11(15)6-4-8-12(9)16-13-7-3-2-5-10(13)14(17)18/h2-8,16H,1H3,(H,17,18)

InChI key

YEZNLOUZAIOMLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tolfenamic acid is a fenamate and is made up of a monocarboxylate diphenylamine nucleus.

Application

Tolfenamic acid has been used in Arabidopsis bud assay for activity comparison with quinazolinedione derivatives (QADD) based compounds.

Biochem/physiol Actions

Non-steroidal anti-inflammatory agent. Interferes with synthesis of β-amyloid precursor protein, and thus Aβ peptides, by promoting degradation of an essential transcription factor.
Tolfenamic acid is involved in the inhibition of prostaglandin synthesis. It also plays a role in apoptosis of head and neck cancer cells.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical synthesis and characterization of a new quinazolinedione competitive antagonist for strigolactone receptors with an unexpected binding mode
Hamiaux C, et al.
The Biochemical Journal, 476(12), 1843-1856 (2019)
Vilmalí López-Mejías et al.
Journal of the American Chemical Society, 131(13), 4554-4555 (2009-04-02)
The nonsteroidal anti-inflammatory drug (NSAID) tolfenamic acid (TA), previously thought to be dimorphic, is demonstrated to have at least five polymorphs. The new forms were uncovered through the emerging screening technique of polymer-induced heteronucleation (PIHn). The presence of conformational changes
R A Tokola et al.
Cephalalgia : an international journal of headache, 4(4), 253-263 (1984-12-01)
Tolfenamic acid is a fenamate which inhibits prostaglandin (PG) biosynthesis and may act as a PG antagonist as well. Caffeine and metoclopramide are used in combination with analgesics and ergotamine in the treatment of migraine attacks, but controlled clinical studies
Tolfenamic acid, metoclopramide, caffeine and their combinations in the treatment of migraine attacks
Tokola R A, et al.
Cephalalgia, 4(4), 253-263 (1984)
Alessandra Mattei et al.
Pharmaceutical research, 29(2), 460-470 (2011-09-01)
Crystallization from solution involves nucleation and growth; growth conditions greatly influence self-association behaviors of solute molecules in these steps, affecting crystal packing of organic molecules. We examined the role of pre-nucleation association to provide insights into the mutual influence between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service