Skip to Content
Merck
All Photos(1)

Documents

SRP6472

Sigma-Aldrich

Transferrin human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

DKFZp781D0156, PRO1557, PRO2086, TF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 76 kDa
observed mol wt 74 kDa by SDS-PAGE (DTT-reduced.)

packaging

pkg of 100 μg
pkg of 20 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TF(7018)

General description

Transferrin is encoded by the TF gene that is mapped to human chromosome 3q22.1. It is a glycoprotein containing homologous iron-binding domains at N-terminal and C-terminal ends. Transferrin has a molecular weight of 80,000 Da and belongs to the transferrin superfamily, the members of which include lactoferrin, melanotransferrin, and ovotransferrin.

Application

To supplement neurosphere culture medium and sphere differentiation medium.

Biochem/physiol Actions

Transferrin is an iron-binding glycoprotein that primarily functions in the transport of plasma iron in vertebrates. It may also have a secondary function of facilitating cell proliferation. The binding of ferric iron to transferrin requires the presence of an anion, that functions as a bridge between the metal and the protein.

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM tris, 100 mM glycine, pH 7.0. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide association study identifies genetic loci associated with iron deficiency.
McLaren CE
PLoS ONE, 6 (2011)
Iron transport and storage proteins.
P Aisen et al.
Annual review of biochemistry, 49, 357-393 (1980-01-01)
Fatty acids activate transcription of the muscle carnitine palmitoyltransferase I gene in cardiac myocytes via the peroxisome proliferator-activated receptor alpha.
Brandt JM
The Journal of Biological Chemistry, 273, 23786-23792 (1998)
Roles of the basic helix-loop-helix genes Hes1 and Hes5 in expansion of neural stem cells of the developing brain.
Ohtsuka T
The Journal of Biological Chemistry, 276, 30467-30474 (2001)
Phylogenetic analyses uncover a novel clade of transferrin in nonmammalian vertebrates.
Mohd-Padil H
Molecular Biology and Evolution, 30, 894-905 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service