Skip to Content
Merck
All Photos(1)

Key Documents

SML3761

Sigma-Aldrich

BRD7586

≥98% (HPLC)

Synonym(s):

3-[(4-Chlorophenyl)sulfonyl]-N-[4-(4-pyridinyl)-2-thiazolyl]propanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14ClN3O3S2
Molecular Weight:
407.89
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

BRD7586 is a cell-permeable, and potent anti-CRISPR agent. BRD7586is a potent and selective inhibitor of SpCas9. It potently inhibits SpCas9 in both HEK293T and U2OS.eGFP.PEST cells without altering its expression, or induction of cytotoxicity. BRD7586 enhanced the specificity of SpCas9. BRD7586 inhibits genome editing at diverse endogenous loci. It is twofold more potent than BRD0539.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A general approach to identify cell-permeable and synthetic anti-CRISPR small molecules
Nature Cell Biology, 24(12), 1766-1775 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service