Skip to Content
Merck
All Photos(1)

Documents

SML3300

Sigma-Aldrich

TB21007

≥98% (HPLC)

Synonym(s):

3-(2-Hydroxyethylthio)-6,6-dimethyl-1-(thiazol-2-yl)-6,7-dihydrobenzo[c]thiophen-4(5H)-one, 6,6-Dimethyl-3-(2-hydroxyethyl)thio-1-(thiazol-2-yl)-6,7-dihydro-2-benzothiophen-4(5H)-one, 6,7-Dihydro-3-[(2-hydroxyethyl)thio]-6,6-dimethyl-1-(2-thiazolyl)benzo[c]thiophen-4(5H)-one, TB 21007, TB-21007

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17NO2S3
CAS Number:
Molecular Weight:
339.50
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2=C(SCCO)SC(C3=NC=CS3)=C2CC(C1)(C)C

InChI

1S/C15H17NO2S3/c1-15(2)7-9-11(10(18)8-15)14(20-6-4-17)21-12(9)13-16-3-5-19-13/h3,5,17H,4,6-8H2,1-2H3

InChI key

QILRYFCEXLFIDS-UHFFFAOYSA-N

Biochem/physiol Actions

TB21007 is a gamma-aminobutyric acid type A receptor (GABAA) alpha 5 (α5) subtype-selective (αβ3γ2 Ki in nM = 1.6/α5, 20/α1, 16/α2, 20/α3) negative allosteric modulator (NAM)/inverse agonist (% inhibition at 100 nM = 51/α5, 21/α1, 1/α2, 3/α3, using L(tk-) fibroblast expressing β3γ2 with respective α subunit). TB21007 enhances cognitive performance in a hippocampal-dependent memory task in vivo (0.3 mg/kg i.p. in rats; delayed ‘matching-to-place′ Morris water maze test) without the convulsant or proconvulsant activity (3 mg/kg i.p. in mice) observed with nonselective inverse agonists.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark S Chambers et al.
Journal of medicinal chemistry, 46(11), 2227-2240 (2003-05-16)
In pursuit of a GABA(A) alpha5-subtype-selective inverse agonist to enhance cognition, a series of 6,7-dihydro-2-benzothiophen-4(5H)-ones has been identified as a novel class of GABA(A) receptor ligands. These thiophenes have higher binding affinity for the GABA(A) alpha5 receptor subtype compared to
K McEown et al.
Neuroscience, 252, 169-177 (2013-08-22)
Temporary neuronal inactivation of the ventral hippocampus with the GABAA agonist muscimol suppresses unconditioned fear behavior (anxiety) but inactivation of the dorsal hippocampus does not. On the other hand, inactivating the dorsal hippocampus disrupts fear memory, while inactivating the ventral
Ming Teng Koh et al.
Neurobiology of aging, 91, 1-4 (2020-04-03)
Numerous aging studies have identified a shift in the excitatory/inhibitory (E/I) balance with heightened hippocampal neural activity associated with age-related memory impairment across species, including rats, monkeys, and humans. Neurobiological investigations directed at the hippocampal formation have demonstrated that unimpaired
Yarim E De la Luz-Cuellar et al.
European journal of pharmacology, 858, 172443-172443 (2019-06-11)
The role of spinal α5 subunit-containing GABAA (α5-GABAA) receptors in chronic pain is controversial. The purpose of this study was to investigate the participation of spinal α5-GABAA receptors in the reserpine-induced pain model. Reserpine administration induced tactile allodynia and muscle
Ming Teng Koh et al.
Neuropharmacology, 64, 145-152 (2012-06-27)
A condition of excess activity in the hippocampal formation is observed in the aging brain and in conditions that confer additional risk during aging for Alzheimer's disease. Compounds that act as positive allosteric modulators at GABA(A) α5 receptors might be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service