Skip to Content
Merck
All Photos(1)

Key Documents

SML2227

Sigma-Aldrich

Emricasan

≥98% (HPLC), powder, pan-caspase inhibitor

Synonym(s):

IDN-6556, N-[2-(1,1-dimethylethyl)phenyl]-2-oxoglycyl-N-[(1S)-1-(carboxymethyl)-2-oxo-3-(2,3,5,6-tetrafluorophenoxy)propyl]-L-Alaninamide, PF-03491390

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27F4N3O7
CAS Number:
Molecular Weight:
569.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Emricasan, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)(C)C1=C(NC(C(N[C@@H](C)C(N[C@@H](CC(O)=O)C(COC2=C(F)C(F)=CC(F)=C2F)=O)=O)=O)=O)C=CC=C1

InChI

1S/C26H27F4N3O7/c1-12(31-24(38)25(39)32-16-8-6-5-7-13(16)26(2,3)4)23(37)33-17(10-19(35)36)18(34)11-40-22-20(29)14(27)9-15(28)21(22)30/h5-9,12,17H,10-11H2,1-4H3,(H,31,38)(H,32,39)(H,33,37)(H,35,36)/t12-,17-/m0/s1

InChI key

SCVHJVCATBPIHN-SJCJKPOMSA-N

Biochem/physiol Actions

Antiapoptotic caspase inhibitor; Pan-Caspase Inhibitor
Emricasan is an antiapoptotic pan-caspase inhibitor. It has been in clinical trials for the treatment of non-alcoholic steatohepatitis (NASH) with advanced fibrosis (scarring) and cirrhosis. Emricasan has also been shown to protect infected astrocytes from ZIKV-induced cell death.
Emricasan, formerly known as IDN-6556, in combination with birinapan is used to treat acute myeloid leukemia (AML). This small molecule irreversible inhibitor is under clinical investigation to reduce hepatic injury and liver fibrosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The caspase-8 inhibitor emricasan combines with the SMAC mimetic birinapant to induce necroptosis and treat acute myeloid leukemia
Brumatti G, et al.
Science Translational Medicine, 8, 339ra69-339ra69 (2016)
Carcinogenicity assessment of the pan-caspase inhibitor, emricasan, in Tg.rasH2 mice
Elbekai RH, et al.
Regulatory Toxicology and Pharmacology, 72, 169-178 (2015)
Haiyan Zhang et al.
Frontiers in cardiovascular medicine, 8, 685434-685434 (2021-08-03)
Dexrazoxane (DXZ) reduces cytotoxicity caused by Doxorubicin (DOX). However, the mechanism of DXZ in ferroptosis and cardiomyopathy remains unclear. This research, therefore, explores the role and mechanism of DXZ in DOX-induced ferroptosis and cardiomyopathy in rats. Kaplan-Meier survival analysis was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service