Skip to Content
Merck
All Photos(1)

Key Documents

SML1553

Sigma-Aldrich

MS049 hydrochloride

≥98% (HPLC)

Synonym(s):

N-Methyl-4-(phenylmethoxy)-1-piperidineethanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O · xHCl
CAS Number:
Molecular Weight:
248.36 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CNCCN(CC1)CCC1OCC2=CC=CC=C2.[H]Cl

InChI

1S/C15H24N2O.ClH/c1-16-9-12-17-10-7-15(8-11-17)18-13-14-5-3-2-4-6-14;/h2-6,15-16H,7-13H2,1H3;1H

InChI key

FJZNVFFTIKWXMA-UHFFFAOYSA-N

Biochem/physiol Actions

MS049 is a potent and selective inhibitor of protein arginine methyltransferases (PRMTs) PRMT 4 and PRMT6. MS049 is active in cells. For full characterization details, please visit the MS049 probe summary on the Structural Genomics Consortium (SGC) website.

MS049N is the negative control for the active probe, MS049. To request a sample of the negative control from the SGC, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

MS049 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service