Skip to Content
Merck
All Photos(1)

Key Documents

SML1535

Sigma-Aldrich

DMH2

≥98% (HPLC)

Synonym(s):

4-[6-[4-[2-(4-Morpholinyl)ethoxy]phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]-quinoline, VU0364849

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25N5O2
CAS Number:
Molecular Weight:
451.52
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear

storage temp.

2-8°C

SMILES string

N1(CCOC2=CC=C(C(C=N3)=CN4C3=C(C5=CC=NC6=C5C=CC=C6)C=N4)C=C2)CCOCC1

InChI

1S/C27H25N5O2/c1-2-4-26-24(3-1)23(9-10-28-26)25-18-30-32-19-21(17-29-27(25)32)20-5-7-22(8-6-20)34-16-13-31-11-14-33-15-12-31/h1-10,17-19H,11-16H2

InChI key

DXLXRNZCYAYUED-UHFFFAOYSA-N

General description

DMH2 is a small molecule, which has ability to inhibit BMP type I receptors. The bone morphogenetic proteins (BMP) signal inhibition further decreases expression of Id proteins in lung cells.

Biochem/physiol Actions

DMH2 is a potent antagonist of the BMP receptor activin receptor-like kinase 3 (ALK3, BMPR-IA) that enhance liver regeneration. DMH2 potently inhibits BMP signaling and induced the greatest reduction of cell growth and expression of Id family members in H1299 non-small cell lung carcinoma cells.
DMH2 is a potent antagonist of the BMP receptor activin receptor-like kinase 3 (ALK3, BMPR-IA).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elaine Langenfeld et al.
Molecular cancer, 12(1), 129-129 (2013-10-29)
Bone morphogenetic proteins (BMP) are embryonic morphogens that are aberrantly expressed in lung cancer. BMPs mediate cell fate decisions and self-renewal of stem cells, through transcription regulation of inhibitor of differentiation protein/DNA binding proteins (Id1-3). Inhibition of BMP signaling decreases
Rui Sun et al.
Cell reports, 36(7), 109559-109559 (2021-08-19)
Acute myeloid leukemia (AML) is a rapidly progressing cancer, for which chemotherapy remains standard treatment and additional therapeutic targets are requisite. Here, we show that AML cells secrete the stem cell growth factor R-spondin 2 (RSPO2) to promote their self-renewal
Fengyu Zhang et al.
Cancer cell international, 20, 197-197 (2020-06-09)
Radiotherapy, chemotherapy, and surgery have made crucial strides in glioblastoma treatment, yet they often fail; thus, new treatment and new detection methods are needed. Aberrant expression of Nanos3 has been functionally associated with various cancers. Here, we sought to identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service