Skip to Content
Merck
All Photos(1)

Key Documents

SML1139

Sigma-Aldrich

Rostafuroxin

≥98% (HPLC)

Synonym(s):

(3,5,14)-21,23-epoxy24-Norchola-20,22-diene-3,14,17-triol, PST 2238

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34O4
CAS Number:
Molecular Weight:
374.51
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

InChI

1S/C23H34O4/c1-20-8-5-17(24)13-15(20)3-4-19-18(20)6-9-21(2)22(25,10-11-23(19,21)26)16-7-12-27-14-16/h7,12,14-15,17-19,24-26H,3-6,8-11,13H2,1-2H3/t15-,17+,18+,19-,20+,21-,22+,23+/m1/s1

InChI key

AEAPORIZZWBIEX-DTBDINHYSA-N

Biochem/physiol Actions

Rostafuroxin is a derivative of digitoxigenin. It protects animal models against podocyte lesions and proteinuria by counteracting mutant β-adducin and ouabain-mediated effects.
Rostafuroxin is a digitoxygenin derivative that selectively antagonizes the effects of endogenous ouabain on the Na+,K+-ATPase, which maintains sodium and potassium ion gradients across plasma membranes. Rostafuroxin lowers blood pressure (BP) in animal models of hypertension.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mara Ferrandi et al.
The Journal of pharmacology and experimental therapeutics, 351(2), 278-287 (2014-09-05)
Glomerulopathies are important causes of morbidity and mortality. Selective therapies that address the underlying mechanisms are still lacking. Recently, two mechanisms, mutant β-adducin and ouabain, have been found to be involved in glomerular podocytopathies and proteinuria through nephrin downregulation. The
David W Sanders et al.
eLife, 10 (2021-04-24)
Many enveloped viruses induce multinucleated cells (syncytia), reflective of membrane fusion events caused by the same machinery that underlies viral entry. These syncytia are thought to facilitate replication and evasion of the host immune response. Here, we report that co-culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service