Skip to Content
Merck
All Photos(1)

Key Documents

SML0834

Sigma-Aldrich

2,3,5,4′-Tetrahydroxystilbene 2-O-β-D-glucoside

≥98% (HPLC)

Synonym(s):

2,4-Dihydroxy-6-[(1E)-2-(4-hydroxyphenyl)ethenyl]phenyl β-D-Glucopyranoside, EH-201, THSG, Tetrahydroxystilbene glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O9
CAS Number:
Molecular Weight:
406.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to brown

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

−20°C

InChI

1S/C20H22O9/c21-9-15-16(25)17(26)18(27)20(28-15)29-19-11(7-13(23)8-14(19)24)4-1-10-2-5-12(22)6-3-10/h1-8,15-18,20-27H,9H2/b4-1+/t15-,16-,17+,18-,20+/m1/s1

InChI key

JAYVHSBYKLLDJC-DSNJPTTOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,5,4′ -Tetrahydroxystilbene-2-O-beta-glucoside (THSG) is a Polygoni Multiflori based polyphenol.

Biochem/physiol Actions

2,3,5,4′-Tetrahydroxystilbene 2-O-β-D-glucoside (THSG) elicits anti-inflammatory, antioxidative and anti-atherosclerotic functionality. It also provides protection against cytotoxicity in neuroblastoma cells induced by 1-methyl-4-phenyl pyridium (MPP+) as well as MPTP (1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine) induced Parkinson′s disease. By its antioxidative functionality, it blocks lipid peroxidation in Alzheimer′s disease model. In ligature-induced periodontitis, THSG inhibits inflammation.
2,3,5,4′-Tetrahydroxystilbene 2-O-β-D-glucoside induces the expression and enhances the activity of EPO. The compound stimulates mitochondrial biogenesis, and expression of hemoglobin in non-hematopoietic cells. 2,3,5,4′-Tetrahydroxystilbene 2-O-β-D-glucoside has demonstrated protective effects in cardiovascular disease and ischemia models.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Hong He et al.
European journal of pharmacology, 767, 175-182 (2015-10-20)
Parkinson's disease (PD) is characterized by the selective death of dopaminergic neurons in the substantia nigra pars compacta. Oxidative stress-induced neuron loss is thought to play a crucial role in the pathogenesis of PD. Previous work from our group suggests
Yu-Tang Chin et al.
Mediators of inflammation, 2016, 6953459-6953459 (2016-08-10)
Periodontitis, a chronic infection by periodontopathic bacteria, induces uncontrolled inflammation, which leads to periodontal tissue destruction. 2,3,5,4'-Tetrahydroxystilbene-2-O-beta-glucoside (THSG), a polyphenol extracted from Polygoni Multiflori, reportedly has anti-inflammatory properties. In this study, we investigated the mechanisms of THSG on the Porphyromonas
Yun-Ho Hwang et al.
International journal of molecular sciences, 19(12) (2018-12-14)
Asthma is an inflammatory disease caused by an imbalance of Th1 and Th2 cells. In general, asthma is characterized by a stronger Th2 response. Most conventional asthma treatment focuses on improving airway flow or suppression of airway inflammation. To reduce

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service