Skip to Content
Merck
All Photos(1)

Key Documents

SML0240

Sigma-Aldrich

Adefovir

≥98% (HPLC)

Synonym(s):

GS 0393, GS-393, P-[[2-(6-Amino-9H-purin-9-yl)ethoxy]methyl]phosphonic acid, 9-(2-phosphonylmethoxyethyl)adenine, PMEA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N5O4P
CAS Number:
Molecular Weight:
273.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

0.1 M NaOH: ≥5 mg/mL

storage temp.

−20°C

SMILES string

Nc1ncnc2n(CCOCP(O)(O)=O)cnc12

InChI

1S/C8H12N5O4P/c9-7-6-8(11-3-10-7)13(4-12-6)1-2-17-5-18(14,15)16/h3-4H,1-2,5H2,(H2,9,10,11)(H2,14,15,16)

InChI key

SUPKOOSCJHTBAH-UHFFFAOYSA-N

Application

Adefovir has been used to study its anti-retro viral effect on porcine endogenous retrovirus (PERV) activity.

Biochem/physiol Actions

Adefovir is an antiviral and is the active form of adefovir dipivoxyl
Adefovir is an antiviral drug that after intracellular conversion to adefovir diphosphate inhibits hepatitis B virus (HBV) DNA polymerase (reverse transcriptase).
Adefovir, also known as 9-(2-phosphonylmethoxyethyl)adenine (PMEA), is an acyclic nucleoside phosphonate. It has a potential to hinder the in vitro replication of several retroviruses such as human immunodeficiency virus (HIV)-1 and HIV-2, simian immunodeficiency virus (SIV), simian AIDS-related virus (SRV), feline immunodeficiency virus (FIV). Thus, Adefovir may be used as a therapeutic for various retrovirus infections including acquired immunodeficiency syndrome (AIDS).

Features and Benefits

This compound is a featured product for ADME Tox and Cyclic Nucleotide research. Discover more featured ADME Tox and Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenylyl cyclases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anders Boyd et al.
Journal of hepatology, 65(4), 683-691 (2016-05-24)
In the presence of highly-potent antivirals, persistence of hepatitis B virus (HBV) is most well-characterized by covalently-closed circular DNA (cccDNA) and total intrahepatic DNA (IH-DNA). We sought to determine how antiviral therapy could affect their levels during human immunodeficiency virus
9-(2-Phosphonylmethoxyethyl) adenine (PMEA) effectively inhibits retrovirus replication in vitro and simian immunodeficiency virus infection in rhesus monkeys.
Balzarini J, et al.
AIDS, 5(1), 21-28 (1991)
Valeria Cento et al.
The Journal of infection, 67(4), 303-312 (2013-06-26)
The identification of novel reverse-transcriptase (RT) drug-resistance mutations is critical in predicting the probability of success to anti-HBV treatment. Furthermore, due to HBV-RT/HBsAg gene-overlap, they can have an impact on HBsAg-detection and quantification. 356 full-length HBV-RT sequences from 197 drug-naive
Gang Huang et al.
Annals of surgery, 261(1), 56-66 (2014-07-30)
A randomized controlled trial was conducted to find out whether antiviral therapy in patients with hepatitis B-related hepatocellular carcinoma (HCC) improves long-term survival after hepatic resection. Despite advances in surgery and in multidisciplinary treatment, there is still no effective adjuvant
Jan Balzarini et al.
PLoS pathogens, 9(7), e1003456-e1003456 (2013-08-13)
Human immunodeficiency virus (HIV) infection is often accompanied by infection with other pathogens, in particular herpes simplex virus type 2 (HSV-2). The resulting coinfection is involved in a vicious circle of mutual facilitations. Therefore, an important task is to develop

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service