Skip to Content
Merck
All Photos(1)

Documents

SML0074

Sigma-Aldrich

Acenocoumarol

≥98% (HPLC)

Synonym(s):

(±)-Acenocoumarin, 3-(α-Acetonyl-p-nitrobenzyl)-4-hydroxy-coumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15NO6
CAS Number:
Molecular Weight:
353.33
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO, heptane and xylene: ≥17 mg/mL

originator

Novartis

storage temp.

−20°C

SMILES string

CC(=O)CC(c1ccc(cc1)[N+]([O-])=O)C2=C(O)c3ccccc3OC2=O

InChI

1S/C19H15NO6/c1-11(21)10-15(12-6-8-13(9-7-12)20(24)25)17-18(22)14-4-2-3-5-16(14)26-19(17)23/h2-9,15,22H,10H2,1H3

InChI key

VABCILAOYCMVPS-UHFFFAOYSA-N

Gene Information

human ... VKORC1(79001)

Looking for similar products? Visit Product Comparison Guide

Application

Acenocoumarol has been used as a standard for the determination of coumarins in cosmetics.
Acenocoumarol was used to study the role of P-glycoprotein in transport of oral vitamin K in Caco-2 cells and as an LC/MS standard.

Biochem/physiol Actions

Acenocoumarol is a warfarin analog, an anticoagulant that inhibits Vitamin K epoxide reductase. This results in depletion of the reduced form of vitamin K (vitamin KH2), limiting the gamma-carboxylation and subsequent activation of the vitamin K-dependent coagulation factors II, VII, IX, and X and anticoagulant proteins C and S, resulting in decreased prothrombin levels and the amount of thrombin generated.
Acenocoumarol is effective against thromboembolic disorders.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service