Skip to Content
Merck
All Photos(1)

Key Documents

SMB01018

Sigma-Aldrich

Cynaropicrin

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O6
CAS Number:
Molecular Weight:
346.37
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

346.37

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@@H]2[C@@H]3[C@@H](C[C@@H](C3=C)O)C(=C)C[C@@H]([C@H]2C(=C)C1=O)OC(=O)C(=C)CO

InChI

1S/C19H22O6/c1-8-5-14(24-18(22)9(2)7-20)16-11(4)19(23)25-17(16)15-10(3)13(21)6-12(8)15/h12-17,20-21H,1-7H2/t12-,13-,14-,15-,16+,17+/m0/s1

InChI key

KHSCYOFDKADJDJ-NQLMQOPMSA-N

General description

Cynaropicrin is a sesquiterpene lactone, a bioactive natural compound commonly derived from plants such as Cynara scolymus, Pleiotaxis rugosa, Pseudostifftia kingii, Blumea eriantha, and Scopolia atropoides. Current research indicates that this plant metabolite is an inhibitor and may possess diverse biological activities, including anti-inflammatory, antineoplastic, antispasmodic, anti-hyperlipidemic, anti-trypanosomal, anti-malarial, and antiviral properties.

Application

Cynaropicrin is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Cynaropicrin is a potent activator of the AhR-Nrf2-Nqo1 pathway, making it a potential candidate for preventing UVB-induced photoaging. Additionally, it exhibits pro-apoptotic activity, suggesting its potential as an anticancer agent against certain leukocyte cancer cells such as lymphoma or leukemia. Cynaropicrin has demonstrated in vivo activity against Trypanosoma brucei and exhibits immunomodulatory effects by influencing cytokine release and nitric oxide production while also displaying immunosuppressive properties. Moreover, it has anti-inflammatory effects by inhibiting the production of inflammatory mediators and lymphocyte proliferation, which is achieved through conjugation with sulphydryl groups of target protein(s).

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service