Skip to Content
Merck
All Photos(5)

Key Documents

SAB4504094

Sigma-Aldrich

Anti-phospho-p38 MAPK (pTyr322) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 41 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr322)

Gene Information

human ... MAPK14(1432)

General description

MAPK14 (mitogen-activated protein kinase 14) is a Ser/Thr kinase made of 90 to 360 residues. p38αMAPK is ubiquitously expressed. It is found in the cytosol, but it can translocate to the nucleus. This gene is located on human chromosome 6p21.31.

Immunogen

The antiserum was produced against synthesized peptide derived from human p38 MAPK around the phosphorylation site of Tyr322.

Immunogen Range: 288-337

Biochem/physiol Actions

MAPK14 (mitogen-activated protein kinase 14) participates in resistance of colon cancer cells to camptothecin-related drugs. This protein is required for the growth of embryos. This gene controls proliferation, differentiation, cell death, adhesion, migration, as well as the response to stress and many metabolic pathways. Polymorphism in MAPK14/p38a leads to deletion or overproduction of myelosuppressive cytokines and result in certain bone marrow failure syndromes.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MAPK14 (mitogen activated protein kinase 14)
Porras A and Guerrero C
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2010)
MAPK14/p38? confers irinotecan resistance to TP53-defective cells by inducing survival autophagy.
Paillas S, et al.
Autophagy, 8(7), 1098-1112 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service