Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500654

Sigma-Aldrich

Anti-SIGLEC15 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

mol wt

36 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Sialic acid binding Ig like lectin 15 (SIGLEC15) is part of the Siglec family of glycan-recognition proteins. It is expressed in a subset of macrophages. This type-I transmembrane protein possesses two immunoglobulin-like domains, a transmembrane domain and a short cytoplasmic tail. The SIGLEC15 gene is localized on human chromosome 18q12.3.

Immunogen

Antibody was raised against a 15 amino acid peptide near the amino terminus of human SIGLEC15.

Biochem/physiol Actions

Sialic acid binding Ig like lectin 15 (SIGLEC15) recognizes a tumor-linked glycan structure called the sialyl-Tn (sTn) antigen. It is expressed on tumor-associated macrophages in tumor tissues. The protein acts as a signal transducer. SIGLEC15 associates with the adaptor protein DNAX activation protein of 12kDa (DAP12) and transduces a signal to spleen tyrosine kinase (Syk).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500654.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun-Long Wang et al.
American journal of cancer research, 11(5), 2291-2302 (2021-06-08)
Siglec15 is a recently characterized immunosuppressive transmembrane protein, which expresses in various types of solid tumors and promotes cancer development. Several studies reported that Siglec15 is a prognostic biomarker of cancer patients, and targeting Siglec15 may be a promising strategy
Kaiqin Sheng et al.
International journal of molecular sciences, 24(1) (2023-01-09)
Sialic acid-binding immunoglobulin-like lectin 15 (Siglec-15) has been identified as a crucial immune suppressor in human cancers, comparable to programmed cell death 1 ligand (PD-L1). However, the regulatory mechanisms underlying its transcriptional upregulation in human cancers remain largely unknown. Here
Siglec-15: an immune system Siglec conserved throughout vertebrate evolution.
Glycobiology (2007)
Xiaoting Chen et al.
Heliyon, 10(3), e25266-e25266 (2024-02-14)
Laryngeal squamous cell carcinoma (LSCC) is the ultimate common malignant head and neck cancer with dismal prognosis. The expression pattern and clinical significance of Siglec-15 (Sialic acid-binding immunoglobulin-like lectin 15) in LSCC are poorly understood. In order to lay the
Comparative genomics indicates the mammalian CD33rSiglec locus evolved by an ancient large-scale inverse duplication and suggests all Siglecs share a common ancestral region
Huan Cao
Immunogenetics, 401-477 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service