Skip to Content
Merck
All Photos(1)

Key Documents

S1820

Sigma-Aldrich

Ser-Phe-Leu-Leu-Arg-Asn-amide trifluoroacetate salt

≥98% (HPLC), lyophilized powder

Synonym(s):

PAR-1 (Protease-Activated Receptor) Selective Activating Peptide, PAR1-AP, SFLLRN-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C34H57N11O8 · x CF3COOH
CAS Number:
Molecular Weight:
747.89 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (HPLC)

form

lyophilized powder

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)CO)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(N)=O)C(N)=O

InChI

1S/C34H57N11O8.C2HF3O2/c1-18(2)13-24(31(51)41-22(11-8-12-40-34(38)39)30(50)42-23(28(37)48)16-27(36)47)44-32(52)25(14-19(3)4)45-33(53)26(43-29(49)21(35)17-46)15-20-9-6-5-7-10-20;3-2(4,5)1(6)7/h5-7,9-10,18-19,21-26,46H,8,11-17,35H2,1-4H3,(H2,36,47)(H2,37,48)(H,41,51)(H,42,50)(H,43,49)(H,44,52)(H,45,53)(H4,38,39,40);(H,6,7)/t21-,22-,23-,24-,25-,26-;/m0./s1

InChI key

YICKZNIVMKPQPD-POCCEKOUSA-N

Gene Information

human ... IGF1R(3480)

Amino Acid Sequence

Ser-Phe-Leu-Leu-Arg-Asn-NH2

Application

Ser-Phe-Leu-Leu-Arg-Asn-amide trifluoroacetate salt has been used as a protease-activated receptor 1 (PAR-1) selective activating peptide.

Biochem/physiol Actions

Ser-Phe-Leu-Leu-Arg-Asn-amide trifluoroacetate salt (SFLLRN) acts as a protease-activated receptor 1 (PAR-1) specific agonist peptide.
Thrombin receptor activating peptide (TRAP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. S. Davies
Amino Acids, Peptides and Proteins, 29 (1998)
Thrombin induces Ca2+-dependent glutamate release from RPE cells mediated by PLC/PKC and reverse Na+/Ca2+ exchange
Lopez E, et al.
Molecular Vision (2019)
Nicola Maggio et al.
Hippocampus, 24(8), 1030-1038 (2014-04-23)
Thrombin and other clotting factors regulate long-term potentiation (LTP) in the hippocampus through the activation of the protease activated receptor 1 (PAR1) and consequent potentiation of N-methyl-d-aspartate receptor (NMDAR) functions. We have recently shown that the activation of PAR1 either
B Batman et al.
Vox sanguinis, 112(8), 773-779 (2017-09-30)
Prophylactic platelet transfusions are administered to prevent bleeding in haemato-oncological patients. However, bleeding still occurs, despite these transfusions. This practice is costly and not without risk. Better predictors of bleeding are needed, and flow cytometric evaluation of platelet function might
Visualizing dynamics of cell signaling in vivo with a phase separation-based kinase reporter
Zhang Q, et al.
Molecular Cell, 69(2), 334-346 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service