Skip to Content
Merck
All Photos(1)

Documents

S1577

Sigma-Aldrich

Stromal Cell-Derived Factor 1α/pre-B Cell Growth Stimulating Factor human

recombinant, expressed in E. coli, lyophilized powder, ≥97% (SDS-PAGE), suitable for cell culture

Synonym(s):

SDF-1, SDF1, SDF-1α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

≤50 ng/mL EC50

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg protein Endotoxin

color

white

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CXCL12((6387)

General description

Stromal Cell-Derived Factor 1α (SDF-1α) or pre-B Cell Growth Stimulation Factor (PBSF) is an 8 kDa chemokine peptide. SDF-1 was identified using signal sequence trap cloning. With this method, cDNAs have been cloned using mouse and human stromal cell lines. This cytokine is widely accepted as a CXC chemokine of the intercrine-macrophage inflammatory protein superfamily. The gene has been mapped to chromosome 10q and is encoded by 4 exons. Expression of the human SDF1 gene is abundant in the pancreas, spleen, ovary, and small intestine. The presence of a GC-rich sequence and the lack of a TATA box in the 5∅-flanking region of the gene are consistent with ubiquitous expression.

Biochem/physiol Actions

SDF-1α functions as a pre-B cell growth factor in the presence of IL-7 (interleukin-7) and a potent chemoattractant for T-lymphocytes and monocytes. It has also been found to be a ligand for CXCR4 (chemokine receptor type 4) or the orphan receptor LESTR/fusin (putative chemokine receptor). By signaling through the receptor, SDF1 may serve as an inhibitor of HIV-1 (human immunodeficiency virus), which utilizes the LESTR/fusin receptor as a point of entry. The SDF-1/CXCR4 signalling pathway is associated with cancer cell proliferation, migration and survival. Overexpression of SDF1 is observed in breast carcinoma, ovarian carcinoma and papillary thyroid carcinoma. It enhances angiogenesis in the ischemic tissue. SDF1 offers CXCR4 mediated neuroprotection during post ischemic stroke.

Physical form

This product is lyophilized from a 0.2 μm-filtered solution containing 10 μg SDF-1α and 0.5 mg bovine serum albumin in PBS.

Reconstitution

Reconstitute the contents of the vial using sterile buffer or medium containing a minimum of 0.1% BSA or HSA to a stock concentration of ≥10 μg/ml. Additional filtration of the stock solution is not recommended as this may result in loss of product due to adsorption onto the filter membrane.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SDF-1/CXCR4 expression is an independent negative prognostic biomarker in patients with head and neck cancer after primary radiochemotherapy
Colle CD, et al.
Radiotherapy and Oncology : Journal of the European Society for Therapeutic Radiology and Oncology, 126, 125-131 (2018)
Chemokines in multiple myeloma
Aggarwal R, et al.
Experimental Hematology, 34(10), 1289?-12295 (2006)
Diversity and inter-connections in the CXCR4 chemokine receptor/ligand family: molecular perspectives
Pawig L, et al.
Frontiers in Immunology (2015)
The α-Chemokine, Stromal Cell-derived Factor-1α, Binds to the
Transmembrane G-protein-coupled CXCR-4 Receptor and
Activates Multiple Signal Transduction Pathways.
Ganju RK, et al.
The Journal of Biological Chemistry, 273, 23169?- 233175 (1998)
Structure and Chromosomal Localization of the Human Stromal Cell-Derived Factor 1 (SDF1) Gene
Shirozu M, et al.
Genomics, 28, 495-500 (1995)

Protocols

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service